Avantor® Hichrom PEEKsil™, In-Line Filter
Catalog # HICHHI-683
Supplier: Avantor
Specifications
- Omschrijving:Fingertight Ti PEEK encased inline filter
- VPE:1
Specifications
About this item
This is a fingertight bioanalytical inline filter, comprising a PEEK holder and a 5 μm titanium PEEK-encased frit.
- Protects the injector from particulates
- Compatible with all HPLC systems
- Replacement 5 μm and 2 μm titanium PEEK-encased frits are available
Placed between the pump and sample injection valve, the in-line filter traps particles released through normal piston seal wear within the pump. It is recommended that the frits are checked frequently and replaced as soon as they contribute to a system back pressure.