Avantor® Hichrom, Tubing Cutters, Stainless Steel
HICHHI-192
: Avantor
- Omschrijving:Stainless steel tubing cutter
- Seq:01
- VPE:1
Stainless steel cutters are specifically designed for ¹/₁₆" stainless steel tubing.
- Smooth and uniform cuts
Stainless steel cutters are specifically designed for ¹/₁₆" stainless steel tubing.