Cleanroom mop, Clino One Way Ultra PW-I
141-1457
- Kleur:wit
- Omschrijving:Clino One Way Ultra PW-I mop, 55 g
- l×b:400×120 mm
- Environmentally Preferable:
- VPE:5
Pre-washed and gamma-irradiated sterile premium disposable mop with maximum square meter output.
- For use on all standard cleanroom and pharmaceutical floors
- 100% Polyester microfiber material
- With backfilling for enormous sqm performance
- Produced in a controlled environment, gamma irradiated
Excellent cleaning performance due to polyester microfibre with special structure for optimum particle absorption. Up to 20 m² area output due to back-feeding. Very high-quality processing suitable for cleanrooms ISO 5, 6, 7, 8, GMP A/B, C, D.