Safety helmet, pheos S-KR
Catalog # UVEX9772.544
Supplier: UVEX
Specifications
- Kleur:blauw
- Omschrijving:Safety helmet with ventilation, uvex pheos S-KR
- VPE:1
Specifications
About this item
This safety helmet is equipped with slots for uvex visors and earmuffs.
- Shorter brim for a wider field of upward vision
- Three variable air vents for maximum ventilation
- Adjustable range for helmet is 520 to 610 mm
- Optimum fit and comfort with six point textile suspension
The suspension harness features a wheel ratchet system for individual adjustment, with the possibility to mount uvex pheos goggles. Helmet is premounted with four point chinstrap and a goggle glip.
Adjustable range: 520 to 610 mm