Visitor coat set
wit, XL
Catalogus # 113-9288
Leverancier: VWR Collection
Ga naar:
Product Details & Documents
The visitor coat set consists of one LDPE white visitor coat, one bouffant cap, one paper face mask and two CPE shoecovers.
- No pocket
- With button and collar
Verpakking: 50 sets per dispenser box, 4 boxes per carton.
Documents
Food Contact, BPA, Phthalates Certificate
Aanvullende productdetails
TDS Visitor Coat Set White XL
Technische fiche
Specificaties
- Confectiemaat:XL
- Kleur:wit
- Geslacht:Uniseks
- Cat. No.:113-9288
- Kledingstuk toepassing:Laboratory
- Sluitingstype:Met knoppen
- Zakken:zonder zakken