Test mixture 2 for apolar capillary columns according to Grob, for GC, ready-to-use, Supelco®
: Merck
Test mixture 2 for apolar capillary columns according to Grob, for GC, ready-to-use, Supelco®
: Merck
Frequently Bought Together
pH buffer solutions
: Mettler - Toledo
VWR® AVS TITRINORM pH-bufferoplossingen in zakjes, 20 °C
: VWR Chemicals
VWR® AVS TITRINORM, Bufferoplossingen, 20 °C
: VWR Chemicals
Buffer solutions
: Mettler - Toledo