Orders LinkContactUsLinkComponent
TKN indicator solution
TKN indicator solution
TKN indicator solution, 50 mL
Catalog # 705-0112
Supplier:   Hach
TKN indicator solution
Catalog # 705-0112
Supplier:  Hach
CAS Number:  
TKN indicator solution
TKN indicator solution
TKN indicator solution, 50 mL
CAS Number:  
Price
24,10 €
Log in for availability
1 * 50 mL
Product Details & Documents
Indicator solution for pH adjustment in total kjeldahl nitrogen determination by the Nessler method.
Packaging: Packed in a self-contained dropping bottle.
Specifications
Specifications
  • VPE:
    50 mL
  • Omschrijving:
    TKN indicator solution
Specifications