LAL-test, Pyrotell®-T, turbidimetric formulation
Catalog # CAPET0051-5
Supplier: Associates of Cape Cod
CAS Number:
Specifications
- VPE:5
- Omschrijving:Pyrotell®-T, 0,001 Eu/ml (pyros kinetix tube reader)
Specifications
About this item
Pyrotell®-T lysate is used to quantify endotoxin in kinetic turbidimetric tests.
Pyrotell-T lysate can be used with the Pyros Kinetix® tube reader and also incubating microplate readers. When used with the pyros kinetix tube reader, Pyrotell-T gives a highly economic, flexible and sensitive LAL assay.
Pyrotell-T lysate can be used for a wide variety of endotoxin tests, ranging from standard water testing to samples requiring high sensitivity, such as intrathecal products and those requiring high dilutions to overcome interference.
Delivery: Pyrotell-T lysate is supplied in individual multi-test vials. Each vial contains sufficient reagent for approximately 96 tests.