Order Entry
Belgium
ContactUsLinkComponent
Supelco® Organic Reference Standard, Acetaldehyde solution
Supelco® Organic Reference Standard, Acetaldehyde solution
Catalog # CRM868095
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, Acetaldehyde solution
Catalog # CRM868095
Supplier:  Merck
CAS Number:  

Specifications

  • Concentratie:
    1000 µg/ml in water
  • VPE:
    1
  • Standaard:
    Certified reference material
  • Omschrijving:
    Acetaldehyde solution
  • Storage conditions:
    2…8 °C
  • Shipping conditions:
    WICE
  • M-Clarity quality level:
    MQ100
  • Vlampunt:
    >110 °C
  • Info VPE:
    Ampule of 100×1,2 ml

Specifications

About this item

  • GHS signal: GHS08