Welding cover spectacles, uvex 9161
Catalog # UVEX9161.145
Supplier: UVEX
Specifications
- Type:UV 400+IR protection level 5, anti-scratch
- Kleur:zwart/groen
- Seq:02
- Beglazing:Grey PC
- VPE:1
Specifications
About this item
Comfortable polycarbonate welding eyewear with unrestricted lateral perception.
- Minimized burn-in of welding sparks
- Pressure-free fit with soft uvex duo-flex frame
- Fully adjustable side arms enable wearers to achieve a customised fit
- With new filter technology with perfect colour perception
- With uvex infradur coating
Certifications: EN 169