Cut resistant gloves, uvex protector chemical NK2725B / NK4025B
Leverancier: UVEX
Certificaten
Over dit artikel
These robust cotton/HPPE/glass safety gloves with special NBR coating meet the highest requirements of mechanical protection. They use multi-layer technology to combine impermeability and optimal cut protection.
- Excellent resistance to grease, mineral oils and many chemicals
- Good wearer comfort
- Good grip in damp, wet and oily areas
Typical applications include: Work with high cut hazard and contact with chemicals, handling heavy-duty tools, sheet metal processing, machinery and tool manufacturing.
Certificeringen: EN ISO 374-1:2016 Type A (JKNOPT), EN 388:2016 (4X44C).
Specificaties
- Catalog No:
- UVEX6053609
- UVEX6053610
- 112-5657
- 112-5658
- Model:
- NK4025B
- NK4025B
- NK2725B
- NK2725B
- Ambidexter/handspecifiek:
- Handspecifiek
- Handspecifiek
- Handspecifiek
- Handspecifiek
- Voor eenmalig gebruik/herbruikbaar:
- Herbruikbaar
- Herbruikbaar
- Herbruikbaar
- Herbruikbaar
- Afwerking:
- Full special NBR coating, rough
- Full special NBR coating, rough
- Full special NBR coating, rough
- Full special NBR coating, rough
- Manchetstijl:
- Lang (Gauntlet)
- Lang (Gauntlet)
- Lang (Gauntlet)
- Lang (Gauntlet)
- Boordlengte:
- 400 mm
- 400 mm
- 270 mm
- 270 mm
- EN-norm:
- EN ISO 374-1:2016 Type A (JKNOPT), EN 388:2016 (4X44C)
- EN ISO 374-1:2016 Type A (JKNOPT), EN 388:2016 (4X44C)
- EN ISO 374-1:2016 Type A (JKNOPT), EN 388:2016 (4X44C)
- EN ISO 374-1:2016 Type A (JKNOPT), EN 388:2016 (4X44C)
Specificaties
Frequently Bought Together