Orders LinkContactUsLinkComponent
ESI nebuliser needle replacement kit
ESI nebuliser needle replacement kit
ESI replacement kit used with ES nebuliser
  HEWLG1958-60137
 :   Agilent
ESI nebuliser needle replacement kit
  HEWLG1958-60137
 :  Agilent
ESI nebuliser needle replacement kit
ESI nebuliser needle replacement kit
ESI replacement kit used with ES nebuliser
380,00 €
1 * 1 ST
The kit contains needles, ferrule and a needle holder.
  • Omschrijving:
    ESI replacement kit used with ES nebuliser