Swab sampling kit
Swab sampling kit, incl. 10 ml buffer (in tube) and swab with PS shaft and polyester head
TSCLTS/5/42
This kit includes 10 ml neutralising buffer in a tube and a sampling swab with PS (breakable) shaft and polyester (dacron) tip, packed in a peel pouch.
- Gamma irradiated (Sterility Assurance Level (SAL) of 10⁻⁶)
- Accurate volumes for serial dilution techniques
Recommended for hygiene and environmental monitoring of equipment or surfaces testing where traces of disinfectants are possibly present. Ideal for maintaining and transporting microbiological samples & eliminating any traces of sanistisers or disinfectants remaining on surfaces.
Excellent recovery of organisms including: TVC, Listeria spp., Staphylococci, Enterobacteriaceae eg Salmonella, E.coli (E coli / ecoli / e. coli), Enterobacter, staphylococcus, klebsiella, E. coli Enterobacter, Enterobacter sakazakii.
- Omschrijving:Swab sampling kit, incl. 10 ml buffer (in tube) and swab with PS shaft and polyester head