Pipettes, P-2, Aquisel
Aquisel pipette, P-2, with rubber stopper
Catalogus # SELE1780225
Leverancier: SELECTA
Ga naar:
Product Details & Documents
Polystyrene pipettes with zero-closure system allows the pipette filling, preventing that the blood exceeds the zero point and also avoiding total or partial loss.
- Pipette filling by pressure till the bottom of the tube
- For erythrocyte sedimentation read E.S.R. Westergren method
- Compatible with conventional tubes of 12×86
- Automatic sealing makes it a disposable product
Verpakking: Cardboard package of 200 units. Supplied in 800 units boxes.
Specificaties
- Omschrijving:Aquisel pipette, P-2, with rubber stopper