Knitted cap, Thinsulate®
Knitted skull cap, blauw
Catalogus # SINGBONTHB
Leverancier: SINGER safety
Ga naar:
Product Details & Documents
Knitted cap with Thinsulate® lining.
- Warm and comfortable
- Thinsulate® consists of very thin 100% polyester fibres (moisture resistant, thermal insulator)
- One size fits all
In cold weather, during your outdoor activities these products will protect you effectively against adverse climatic conditions. Also useful for indoor operations in cold environments and unheated rooms. Construction, agriculture, horticulture, fishing, landscape maintenance, driver, transportation, entertainment.
Certificeringen: EU 2016/425 (PPE), cat. I
Bestelinformatie:
Levering:
Verpakking: Carton of 100 pieces and a bundle of 10 pieces.
Let op:
Specificaties
- Type:Knitted skull cap
- Kleur:blauw
- Geslacht:Uniseks
- Cat. No.:SINGBONTHB
- Kledingstuk toepassing:General Purpose
- Risk area:Cold