Automated nucleic acid extraction system, QIAcube
Catalogus # QIAG990395
Leverancier: QIAGEN
Specificaties
- Omschrijving:Starter pack
- VPE:1
Specificaties
Over dit artikel
QIAcube system is a robotic workstation for automated purification of DNA, RNA, or proteins using QIAGEN spin column kits.
Levering: System includes installation and training, one year warranty on parts and labour. Starter pack includes: Reagent bottle racks (3); rack labeling strips (8); 200 µl filter tips (1024); 1000 µl filter tips (2048); 30 ml reagent bottles (18); rotor adapters (240); rotor adapter holder; 1,5 ml elution tubes (240).