Draining rack
Drying rack, height 105 mm
Catalogus # HECH41238010
Leverancier: GLASWARENFABRIK KARL HECHT
Ga naar:
Product Details & Documents
Drying rack made of Plexiglas®, for 12 microscope slides.
The microscope slides are placed vertically on replaceable blotting paper. Two packs of paper can be stored in the base of the drying rack.
Levering: Supplied with two packs of paper.
Specificaties
- Omschrijving:Drying rack, height 105 mm