Sample tubes, PP
12 mm, blauw, Sample tubes, PP, for FC 500 and EPICS™ XL™ flow cytometers, PP, 75 mm
Catalog # BECL2523749
Marketsource
Supplier: Beckman Coulter
Jump to:
Product Details & Documents
Sample tubes for flow cytometer.
Specifications
- Ø inw.:75 mm
- Kleur:blauw
- Omschrijving:Sample tubes, PP, for FC 500 and EPICS™ XL™ flow cytometers
- Lengte:12 mm
- Materiaal:PP