Orders LinkContactUsLinkComponent
Standard mixture stock solution (2 components)
Standard mixture stock solution (2 components)
Standard mixture stock solution, 5000 μg/ml each in cyclohexane, 1 ml/ampoule, 1 mL
Catalogus # REST31630
Leverancier:   Restek
Standard mixture stock solution (2 components)
Catalogus # REST31630
Leverancier:  Restek
CAS-nummer:  
Standard mixture stock solution (2 components)
Standard mixture stock solution (2 components)
Standard mixture stock solution, 5000 μg/ml each in cyclohexane, 1 ml/ampoule, 1 mL
CAS-nummer:  
Price
70,60 €
Log in voor beschikbaarheid
1 * 1 mL
Product Details & Documents
For GC analysis of total petroleum hydrocarbons (TPH) in water.
  • Calibration standard available as diesel #2/motor oil and diesel #2/mineral oil
The mix consists of diesel #2 (additive-free) and mineral oil (additive-free [ USP grade] bp 325 - 460 or C18 - C32 retention time range).
Certificeringen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Specificaties
Specificaties
  • VPE:
    1 mL
  • Omschrijving:
    Standard mixture stock solution, 5000 μg/ml each in cyclohexane
  • Info VPE:
    1 ml/ampoule