Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
1,2-Dichloroethane

1,2-Dichloroethane

1,2-Dichloroethane, 2000 μg/ml in P and T methanol, 1
Catalog # REST30277
Supplier:   Restek
1,2-Dichloroethane
Catalog # REST30277
Supplier:  Restek
CAS Number:  
1,2-Dichloroethane
1,2-Dichloroethane

1,2-Dichloroethane, 2000 μg/ml in P and T methanol, 1

CAS Number:  
Price
54,60 €
Log in for availability
1 * 1 ST
Product Details & Documents

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.

Specifications

Specifications

  • Concentratie:
    2000 μg/ml in P and T methanol
  • VPE:
    1
  • Omschrijving:
    1,2-Dichloroethane
  • Cat. No.:
    REST30277
  • Formule:
    C₂H₄Cl₂
  • CAS No.:
    107-06-2

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.