Orders LinkContactUsLinkComponent
Alaska UST method AK101AA
Alaska UST method AK101AA
Alaska UST Method AK101AA (14 components), 2,000 µg/mL each in P&T methanol,, 1 mL
Catalogus # REST30461
Leverancier:   Restek
Alaska UST method AK101AA
Catalogus # REST30461
Leverancier:  Restek
CAS-nummer:  
Alaska UST method AK101AA
Alaska UST method AK101AA
Alaska UST Method AK101AA (14 components), 2,000 µg/mL each in P&T methanol,, 1 mL
CAS-nummer:  
Price
74,70 €
Log in voor beschikbaarheid
1 * 1 mL
Product Details & Documents
The alaska UST method consists of 14 components they are benzene, ethylbenzene, 1-ethyl-2-methylbenzene, 1-ethyl-3-methylbenzene, 1-ethyl-4-methylbenzene, isopropylbenzene (cumene), n-propylbenzene, toluene, 1,2,3-trimethylbenzene, 1,2,4-trimethylbenzene, 1,3,5-trimethylbenzene, m-xylene, o-xylene and p-xylene.
Certificeringen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Specificaties
Specificaties
  • Concentratie:
    2,000 µg/mL each in P&T methanol,
  • VPE:
    1 mL
  • Omschrijving:
    Alaska UST Method AK101AA (14 components)