trans-Chlordane
trans-Chlordane, 1,000 µg/ml in methanol, 1 ml/ampoule, 1
Catalogus # REST32227
Leverancier: Restek
Ga naar:
Product Details & Documents
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Specificaties
Specificaties
- Concentratie:1,000 µg/ml in methanol
- VPE:1
- Omschrijving:trans-Chlordane
- Info VPE:1 ml/ampoule
- Cat. No.:REST32227
- Omschrijving:trans-Chlordane
- Formule:C₁₀H₆Cl₈
- CAS No.:5103-74-2