Orders LinkContactUsLinkComponent
Hydrometer jar
Hydrometer jar
40 mm, 400 mm
Catalog # GERI0921
Supplier:   GERING
Hydrometer jar
Catalog # GERI0921
Supplier:  GERING
Hydrometer jar
Hydrometer jar
40 mm, 400 mm
Price
74,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Hydrometer cylinder made of glass.
Specifications
  • Inw. hoogte:
    400 mm
  • I-Ø:
    40 mm