Double bosshead, epoxy coated
Catalog # JUCHVL04822
Supplier: JUCHHEIM Laborgerate GmbH
Specifications
- Omschrijving:Double bosshead with M10 brass T-bar bolt
- Materiaal:Smeedbaar ijzer
- VPE:1
- Voor staaf Ø:30 mm
Specifications
About this item
Double bossheads made of malleable iron, epoxy coated.
Certifications: DIN 12895