Wrenches
Wrench for jerrycan closures, DIN 51, 5 l / 10 l, PE
Catalogus # DECO60025
Leverancier: Borer Chemie
Ga naar:
Product Details & Documents
These wrenches are used for jerrycan closures.
Specificaties
- Omschrijving:Wrench for jerrycan closures, DIN 51, 5 l / 10 l
- Materiaal:PE
- Seq:02