ContactUsLinkComponent
Human Recombinant MIF (from E. coli)
undefined
Human Recombinant MIF (from E. coli)
  • Conjugatie:
    Onconjugeerd
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Bron:
    E. coli
  • Species:
    Human
  • Storage conditions:
    −20 °C
  • Endotoxinegehalte:
    Endotoxin LAL (EU/µg) ≤1
  • Biologische activiteiten:
    Bioactivity: No biological activity data is available at this time
  • Gen-ID:
    P14174
  • Reconstitution instructions:
    Sterile water at 0,1 mg/ml
  • Endotoxinevrij:
    N
  • Dragervrij:
    Y
  • Protease-free:
    N
  • Diervrij:
    N
  • UniProtKB:
    P14174
  • Protein/peptide name:
    MIF
  • Zuiverheid:
    ≥95%
  • Moleculair gewicht:
    12,5 kDa
  • Sequentie:
    MPMFIVNTNV PRASVPDGFL SELTQQLAQA TGKPPQYIAV HVVPDQLMAF GGSSEPCALC SLHSIGKIGG AQNRSYSKLL CGLLAERLRI SPDRVYINYY DMNAANVGWN NSTFA
  • Formulering:
    10 mM sodium phosphate, pH 7,5
  • Vrij van nucleasen:
    N
  • Nr:
    RUO
  • Shipping temperature:
    RT
Frequently Bought Together