Sterile tubing welder, SCD® IIB
Catalog # BCTE3NC-C986
Supplier: TERUMO BCT
Specifications
- Omschrijving:Sterile tubing welder, SCD IIB
- VPE:1
Specifications
About this item
The SCD IIB welder creates strong, sterile tubing connections in about 30 seconds.
- Compact and portable
- Ideal for polyvinyl chloride (PVC), C-FLEX® tubing type
- Simple and easy to use and requires no software
- Create both wet-to-dry and dry-to-dry connections with a single welder
SCD IIB welder allows you to perform a sterile connection to transfer, sample or replace components, all while maintaining a functionally closed system in open labs and production areas. Designed for use with 5,5 mm outside diameter tubing with a wall thickness of 8 mm.