Orders LinkContactUsLinkComponent
Power selector, WRT00116
Power selector, WRT00116
Leverancier:   WINKLER
Power selector, WRT00116
Leverancier:  WINKLER
Certificaten
Power selector, WRT00116
1 Opties Hieronder Beschikbaar
Product Details & Documents
Plug-in power selector made of PS with switching mechanism.
  • Rotary switch with bimetal element
  • Signal light for controlling status indication
  • Cable with grounding plug type
Specifications for Product Family
Specificaties
  • Catalog No:
  • WINKWRT00116-XX013
  • WINKWRT00116-XX0CH
  • WINKWRT00116-XX0UK
  • Model:
  • WRT00116
  • WRT00116
  • WRT00116
  • Elektrisch:
  • 230 V; 13 A (ohmic load); 50 Hz
  • 230 V; 13 A (ohmic load); 50 Hz
  • 230 V; 13 A (ohmic load); 50 Hz
  • b×d×h:
  • 120×56×57 mm
  • 120×56×57 mm
  • 120×56×57 mm
Specificaties
Product Family Options
Productinformatie
  • Model
    Beschikbaarheid
    Price
  • WINKWRT00116-XX013
    Power selector
    Power selector
    WRT00116
    Log in voor beschikbaarheid
    Log in voor beschikbaarheid
    1 * 1 ST
    1 * 1 ST - 117,00 €
    117,00 €
    1 * 1 ST
       117,00 €
    Specificaties:
    More
    Specifications
    Cat. No.
    WINKWRT00116-XX013
    Model
    WRT00116
    Elektrisch
    230 V; 13 A (ohmic load); 50 Hz
    b×d×h
    120×56×57 mm
  • WINKWRT00116-XX0CH
    Power selector, CH-plug
    Power selector, CH-plug
    WRT00116
    Discontinued
    Specificaties:
    More
    Specifications
    Cat. No.
    WINKWRT00116-XX0CH
    Model
    WRT00116
    Elektrisch
    230 V; 13 A (ohmic load); 50 Hz
    b×d×h
    120×56×57 mm
  • WINKWRT00116-XX0UK
    Power selector, UK-plug
    Power selector, UK-plug
    WRT00116
    Discontinued
    Specificaties:
    More
    Specifications
    Cat. No.
    WINKWRT00116-XX0UK
    Model
    WRT00116
    Elektrisch
    230 V; 13 A (ohmic load); 50 Hz
    b×d×h
    120×56×57 mm