Order Entry
Belgium
ContactUsLinkComponent
Tributylfosfaat
Tributylfosfaat
Catalog # EHERC17668000
Supplier:  EHRENSTORFER
CAS Number:  
undefined
Tributylfosfaat
Catalog # EHERC17668000
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Verpakking:
    Glass bottle for solids
  • VPE:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :266.32 g/mol
Merck Index:13,09692
Boiling Pt:289 °C (1013 hPa)
Melting Pt:–79 °C
Density:0.982 g/cm³ (20 °C)
Flash Pt:151 °C
EINECS:204-800-2
 :GHS08
 :GHS07
Storage Temperature:Kamertemperatuur
CAS Number:126-73-8
Formula:(CH₃(CH₂)₃O)₃PO
MDL Number:MFCD00009436