Screw cap, without dropper
Screw cap, GL 18, wit, PP / copolymeerhars
Catalog # MUEL14.016.19
Supplier: MUELLER KREMPEL
Jump to:
Product Details & Documents
Self-sealing caps without drip insert, suitable for steam sterilization.
Specifications
- Schroefdraad:GL 18
- Kleur:wit
- Omschrijving:Screw cap
- Materiaal:PP / copolymeerhars