Glass beads
Chemically resistant glass balls suitable for use as mixing beads, boiling stones, or packing for distillation columns.
Chemically resistant glass balls suitable for use as mixing beads, boiling stones, or packing for distillation columns.
Product Family Options
- Ø