2-Mercaptoethanol ≥98.0%
Catalog # 441438F
Supplier: VWR Chemicals
CAS Number:
Specifications
- Verpakking:Glass bottle
- VPE:25 mL
- Seq:2315
Specifications
Specification Test Results
- Assay (GC, area%):98.0 min.
- Density (20/4):1.115 - 1.116
- Identity (IR):Passes test
Specification Test Results
About this item
:
Merck Index:13,05894
Boiling Pt:157 °C (1013 hPa)
Melting Pt:–100 °C
Density:1.12 g/cm³ (20 °C)
Flash Pt:68 °C
ADR:6.1,II
:M-Clarity™ quality level = MQ200
:78.14 g/mol
EINECS:200-464-6
UN:2966
:GHS06
:GHS05
:GHS09
Storage Temperature:Kamertemperatuur
CAS Number:60-24-2
Formula:HSCH₂CH₂OH
MDL Number:MFCD00004890