Potassium acetate
25 kg
Catalogus # LOHM107021.9025
Leverancier: LOHMANN
Ga naar:
Product Details & Documents
Synoniemen: Acetic acid potassium salt
- CAS-nummer:127-08-2
- Formule:CH₃COOK
- MDL-nummer:MFCD00012458
- Molecular Weight:98.14 g/mol
- Merck index:13,07687
- Smelten Pt:304 °C
- Dichtheid:1,57 g/cm³ (25 °C)
- Flits Pt:>250 °C
- Temperatuur bij opslag:Kamertemperatuur
- EINECS:204-822-2
Specificaties
Specificaties
- VPE:25 kg
Specificaties
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



