Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
D-Norvaline

D-Norvaline

Glass bottle for solids, 5 g
Catalogus # MOLEM82229985
Marketsource
Leverancier:   Molekula
D-Norvaline
Catalogus # MOLEM82229985
Leverancier:  Molekula
Marketsource
CAS-nummer:  2013-12-9
D-Norvaline

Glass bottle for solids, 5 g

CAS-nummer:  2013-12-9
Price
Informeer naar de prijs
Log in voor beschikbaarheid
1 * 5 g
Product Details & Documents
Synoniemen:  H-D-Nva-OH, (R)-(-)-Norvaline, D(-)-Norvaline
  • CAS-nummer:2013-12-9
  • Formule:C₅H₁₁NO₂
  • MDL-nummer:MFCD00008097
  • Molecular Weight:117.15 g/mol
  • EINECS:217-936-2
Specificaties

Specificaties

  • Verpakking:
    Glass bottle for solids
  • VPE:
    5 g

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.