ContactUsLinkComponent
Calcium sulphate dihydrate, EMPROVE® ESSENTIAL Ph. Eur., BP, E516 suitable for use as excipient, precipitated, SAFC®
Calcium sulphate dihydrate, EMPROVE® ESSENTIAL Ph. Eur., BP, E516 suitable for use as excipient, precipitated, SAFC®
Catalogus # 1.02160.9050
CAS-nummer:  
Calcium sulphate dihydrate, EMPROVE® ESSENTIAL Ph. Eur., BP, E516 suitable for use as excipient, precipitated, SAFC®
Catalogus # 1.02160.9050
CAS-nummer:  
Specificaties
  • Verpakking:
    Cardboard carton
  • VPE:
    50 kg
  • Seq:
    4418
  • Environmentally Preferable:
Specificaties
Over dit artikel
Synoniemen: Plaster of Paris
 :
 :172.17 g/mol
Merck index:13,01711
Dichtheid:2.32 g/cm³ (20 °C)
Temperatuur bij opslag:Kamertemperatuur
EINECS:231-900-3
CAS-nummer:10101-41-4
Formule:CaSO₄·2H₂O
MDL-nummer:MFCD00149625