Kaliumhydroxide 1 mol/l (1 N) in ethanol, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
Storage Temperature:Kamertemperatuur
EINECS:215-181-3
UN:2924
ADR:3 (8),II
:GHS02
:GHS05
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):0.998 - 1.002 mol/l
- NIST traceable:Confirmed
Specification Test Results
Related Information
Certificates
Search by Lot Number500 ml Glass bottle for liquids DIN 45 (ZPACE8009063)
Packaging Information
spec_VWRC31306.267 spec_VWRC31306.267
Product specification
VWR® Volumetric Solutions for Titration
Product Literature
Frequently Bought Together