Kaliumhydroxide 0.05 mol/l (0.05 N) in ethanol, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
Storage Temperature:Kamertemperatuur
EINECS:215-181-3
UN:1170
ADR:3,II
:GHS02
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):0.0499 - 0.0501 mol/l
- NIST traceable:Confirmed
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101)
Packaging Information
spec_VWRC31305.297
Product specification
VWR® Volumetric Solutions for Titration
Product Literature
Frequently Bought Together