Styreen stabilised, Supelco®
Glass bottle for solids, 250 mg
Supplier: Merck
Jump to:
Product Details & Documents
Synonyms: Phenylethene, Phenylethylene, Vinylbenzene
- CAS Number:100-42-5
- Formula:C₈H₈
- MDL Number:MFCD00008612
- Molecular Weight:104.15 g/mol
- Merck Index:13,08944
- Boiling Pt:145,2 °C (1013 hPa)
- Melting Pt:–31 °C
- Density:0,906 g/cm³ (20 °C)
- Flash Pt:31 °C
- UN:2055
- ADR:3,III
- :M-Clarity™ quality level = MQ100
- EINECS:202-851-5
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Verpakking:Glass bottle for solids
- VPE:250 mg