ContactUsLinkComponent
Tween® 20 (Polysorbate) 10% in aqueous solution peroxide-free
Tween® 20 (Polysorbate) 10% in aqueous solution peroxide-free
  A1284.0100
 :  
Tween® 20 (Polysorbate) 10% in aqueous solution peroxide-free
  A1284.0100
 :  
  • Verpakking:
    Glass bottle
  • VPE:
    100 mL
  • Seq:
    2440
  • Peroxide (as H2O2):
    max. 0.0001 %
 :
 :1227.53 g/mol
 :Koelkast
 :500-018-3
 :9005-64-5
 :C₅₈H₁₁₄O₂₆
 :MFCD00165986