Orders LinkContactUsLinkComponent
Comet SCGE assay kit
Comet SCGE assay kit
Comet SCGE assay kit, 50 Tests
Catalogus # ENZOADI900166
Leverancier:   Enzo Life Sciences
Comet SCGE assay kit
Catalogus # ENZOADI900166
Leverancier:  Enzo Life Sciences
CAS-nummer:  
Comet SCGE assay kit
Comet SCGE assay kit, 50 Tests
CAS-nummer:  
Price
435,00 €
Log in voor beschikbaarheid
1 * 50 Tests
Product Details & Documents
COMET SCGE Assay measures DNA damage by fluorescently detecting the integrity of DNA liberated from cells embedded in low melting point agarose. Upon electrophoresis, fragmented DNA produces a characteristic "comet" shaped tail as small DNA fragments migrate in the gel more rapidly than in-tact genomic DNA.
  • Comet Slides allow direct application of sample without pretreatment
  • Higher throughput sample analysis
  • Hydrophobic barrier allows sample treatment with DNA repair enzymes
For the measurement of DNA Damage, DNA Repair, and Apoptosis in cells and tissue from any species via electrophoresis.
The COMET SCGE Assay is a fast and simple electrophoresis method to detect and quantitate DNA fragmentation in cells associated with DNA damage and apoptosis. A unique nucleic acid stain provides improved sensitivity for DNA visualization compared to ethidium bromide.
Levering: Kit includes lysis solution, comet LM agarose, comet slide, EDTA, CYGREEN® nucleic acid dye.
Specificaties
Specificaties
  • VPE:
    50 Tests
  • Omschrijving:
    Comet SCGE assay kit
  • Cat. No.:
    ENZOADI900166
  • Maat:
    50 tests
  • Opslag condities:
    Ambient
Specificaties
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.