Peptide standard mixture, MassPREP™
Peptide standard mixture, MassPREP™, 5
Catalogus # WATE186002338
Marketsource
Leverancier: WATERS
Peptide standard mixture, MassPREP™
Catalogus # WATE186002338
Leverancier: WATERS
Marketsource
CAS-nummer:
Ga naar:
Product Details & Documents
The MassPREP™ peptide standard mixture is useful for the evaluation of HPLC columns and systems dedicated for peptide separations.
Each standard contains a void volume marker and nine, carefully selected peptides with a broad range of polarities and isoelectric points. All peaks are easily resolved under reversed-phase HPLC conditions typically used for peptide separations.
Specificaties
Specificaties
- VPE:5
- Omschrijving:Peptide standard mixture, MassPREP™