Immersion oil
APPCA0699.0100
:
- VPE:100 mL
- Omschrijving:Immersion oil
- Seq:01
This fluorescence-free immersion oil for microscopy is highly UV-permeable and free of PCB's.
This fluorescence-free immersion oil for microscopy is highly UV-permeable and free of PCB's.