PCR purification kit, agencourt AMPure XP
Leverancier: Beckman Coulter
Certificaten
Over dit artikel
The Agencourt AMPure XP system is a highly efficient, easily automated PCR purification system that delivers superior quality DNA with no salt carryover.
- High recovery of amplicons, greater than 100 bp
- Efficient removal of unincorporated dNTPs, primers, primer dimers, salts and other contaminants
- No PCR degradation after storage at 4 °C for seven days
- Efficient recovery of double stranded and single stranded DNA templates
- Consistent recovery throughout the kit’s 12 month shelf life
- Faster manual and automated processing as compared to traditional post-PCR cleanup methods
Requires no centrifugation or filtration step, Agencourt AMPure XP can be easily used in manual and automated 96- and 384-well formats.
Frequently Bought Together