Knitted gloves, GL13
Leverancier: Portwest
Certificaten
Over dit artikel
These acrylic knitted gloves are lined with an insulating and comfortable Insulatex™ fleece.
- Durable knit
- Soft lining for great levels of warmth
- Universal size
Ideal for leisure or work in low risk environments.
Specificaties
- Catalog No:
- PRTWGL13BKR
- PRTWGL13NAR
- Model:
- GL13
- GL13
- Ambidexter/handspecifiek:
- Handspecifiek
- Handspecifiek
- Voor eenmalig gebruik/herbruikbaar:
- Herbruikbaar
- Herbruikbaar
- Voeringmateriaal:
- Insulatex™
- Insulatex™
- Steken per inch (Gauge):
- 9
- 9
- Manchetstijl:
- Gebreide manchet
- Gebreide manchet
Specificaties
Frequently Bought Together