Orders LinkContactUsLinkComponent
Distribution head
Distribution head
Distribution head, glass for VAPODEST
Catalogus # GERH12-0363
Leverancier:   GERHARDT
Distribution head
Catalogus # GERH12-0363
Leverancier:  GERHARDT
Distribution head
Distribution head, glass for VAPODEST
Price
465,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
This distribution head is made from glass. It prevents that any sample residue gets into the receiver.
Specificaties
  • Omschrijving:
    Distribution head, glass for VAPODEST