pH electrode
Combined pH/temp electrode, with NTC 30 kOhm, Type 230, 0 - 14, Kunststof, Gel, BNC
Catalog # LOVI721230
Supplier: Lovibond
Jump to:
Product Details & Documents
Combined pH/temperature electrode with temperature sensor NTC 30 kOhm and BNC connection.
Specifications
- Aansluiting:BNC
- Type:Type 230
- Omschrijving:Combined pH/temp electrode, with NTC 30 kOhm
- Materiaal:Kunststof
- Elektroliet:Gel
- pH:0 - 14