Deuterium lamp
Deuterium lamp, D2, 35 W
Catalog # KNAUA4071
Supplier: KNAUER
Jump to:
Product Details & Documents
This deuterium lamp is suitable for K-2501, P-311 and P-314 photometers.
Specifications
- Omschrijving:Deuterium lamp, D2, 35 W