Halogen lamp
Narva halogen lamp, 6 V 25 W, with base PY16-1,25, luminous flux 510 lm, life 100 h, filament of 1,7×1,7 mm, Medical and scientific applications, 25 W
Catalogus # TSSP55147
Leverancier: TECHNICAL SERVICE SPARE PARTS
Ga naar:
Product Details & Documents
Narva halogen special lamps are specifically designed for special lighting tasks. These lamps with prefocus base are suitable for scientific-technical equipment due to their design and optical precision.
- Made of quartz glass
- Transmits UV radiation
- Operating position of s90
- With prefocus base and mounting plate
Applications: Microscopy, ophthalmology, dermatology, food analysis, blood analysis, illumination for surgeries and analysis, measurement technology, projection.
Specificaties
- Voor:Medical and scientific applications
- Omschrijving:Narva halogen lamp, 6 V 25 W, with base PY16-1,25, luminous flux 510 lm, life 100 h, filament of 1,7×1,7 mm
- Vermogen:25 W
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.