Orders LinkContactUsLinkComponent
Spray chamber
Spray chamber
Spray chamber
Catalog # HEWLG3139-65003
Supplier:   Agilent
Spray chamber
Catalog # HEWLG3139-65003
Supplier:  Agilent
Spray chamber
Spray chamber
Price
3 810,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This PFA spray chamber is the standard spray chamber used with the 7500 ICP-MS when aspirating hydrofluoric acid.
Specifications
  • Omschrijving:
    Spray chamber