Orders LinkContactUsLinkComponent
Acetate buffer solution, Fluka™
Acetate buffer solution, Fluka™
Acetate buffer solution, Fluka™
Acetate buffer solution, Fluka™
This pH buffer solution is a combination of sodium acetate/acetic acid in 1:1 ratio for complexometry.
Documents
Product Family Options
  • Omschrijving
    pH-waarde
    Verpakking
    VPE
  • Acetate buffer solution
    4,6
    Plastic bottle
    500 mL
    1 * 500 mL
    1 * 500 mL - 49,60 €
    49,60 €
    1 * 500 mL
       49,60 €
  • Acetate buffer solution
    4,6
    Plastic bottle
    1 L
    1 * 1 L
    1 * 1 L - 62,40 €
    62,40 €
    1 * 1 L
       62,40 €
  • Acetate buffer solution
    4,65
    Plastic bottle
    1 L
    1 * 1 L
    1 * 1 L - 60,00 €
    60,00 €
    1 * 1 L
       60,00 €
  • Acetate buffer solution
    4,65
    Plastic bottle
    6 L
    1 * 6 L
    1 * 6 L - 324,00 €
    324,00 €
    1 * 6 L
       324,00 €